2,6,10-Trimethyl-2,6,10-triazaundecane ≥95.0%

Supplier: TCI America

Synonyms: N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine

T1618-25ML T1618-500ML
TCT1618-025MLEA 36 USD
TCT1618-025ML TCT1618-500ML
2,6,10-Trimethyl-2,6,10-triazaundecane ≥95.0%
2,6,10-Trimethyl-2,6,10-triazaundecane

CAS Number: 3855-32-1
MDL Number: MFCD00126936
Molecular Formula: C11H27N3
Molecular Weight: 201.36
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 102
Flash Point (°C): 92
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine

Formula: C₁₁H₂₇N₃
MW: 201.36 g/mol
Boiling Pt: 102 °C
Density: 0.83 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00126936
CAS Number: 3855-32-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2998e4fe35776720860cd74f2f8a4464b1823ce&vid=b2998e4fe35776720860cd74f2f8a4464b1823ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR