Triphenylsulfonium tetrafluoroborate ≥98.0% (by HPLC)

Supplier: TCI America

T1608-1G T1608-5G
TCT1608-001GEA 113 USD
TCT1608-001G TCT1608-5G
Triphenylsulfonium tetrafluoroborate ≥98.0% (by HPLC)
Triphenylsulfonium tetrafluoroborate

CAS Number: 437-13-8
MDL Number: MFCD00671581
Molecular Formula: C18H15BF4S
Molecular Weight: 350.18
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 194

Formula: C₁₈H₁₅BF₄S
Melting Pt: 194 °C
MDL Number: MFCD00671581
CAS Number: 437-13-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a79db1f1188c473f2fc9c30a03ed25ac5481ca73&vid=a79db1f1188c473f2fc9c30a03ed25ac5481ca73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR