2,2,2-Trifluoroethyl trifluoromethanesulfonate ≥95.0%

Supplier: TCI America

Synonyms: 2,2,2-Trifluoroethyltrifluoromethanesulphonate

T1604-5G T1604-25G
TCT1604-005GEA 143 USD
TCT1604-005G TCT1604-25G
2,2,2-Trifluoroethyl trifluoromethanesulfonate ≥95.0%
2,2,2-Trifluoroethyl trifluoromethanesulphonate

CAS Number: 6226-25-1
MDL Number: MFCD00671579
Molecular Formula: C3H2F6O3S
Molecular Weight: 232.10
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 91
Specific Gravity (20/20): 1.61


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Fluorinated Building Blocks, Other Fluorinated Group Introducing Agents
Other Product Names / Synonyms: 2,2,2-Trifluoroethyl Triflate, Trifluoromethanesulfonic Acid 2,2,2-Trifluoroethyl Ester

Formula: C₃H₂F₆O₃S
MW: 232.10 g/mol
Boiling Pt: 91 °C
Density: 1.61 (20 °C)
MDL Number: MFCD00671579
CAS Number: 6226-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a702fcb09694f2965e23cd918ee9580c0cf70b22&vid=a702fcb09694f2965e23cd918ee9580c0cf70b22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR