1,4,7-Triazacyclononane trihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
1,4,7-Triazonane trihydrochloride
T1600-1G
T1600-5G
TCT1600-001GEA
140
USD
TCT1600-001G
TCT1600-005G
1,4,7-Triazacyclononane trihydrochloride ≥98.0% (by total nitrogen and titration analysis)
1,4,7-Triazacyclononane trihydrochloride
CAS Number: 58966-93-1
MDL Number: MFCD00074887
Molecular Formula: C6H15N3
Molecular Weight: 238.58
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Flash Point (°C): 110
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Polyamines
Other Product Names / Synonyms: Octahydro-1H-1,4,7-triazonine Trihydrochloride
Formula:
C₆H₁₈Cl₃N₃ MW: 238.5900 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD00074887 CAS Number: 58966-93-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (N,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...