2,4,6-Trimethoxyboroxin ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Trimethoxycyclotriboroxane, Trimethoxyboroxine
T1581-25G
T1581-100G
TCT1581-025GEA
24
USD
TCT1581-025G
TCT1581-100G
2,4,6-Trimethoxyboroxin ≥95.0% (by titrimetric analysis)
2,4,6-Trimethoxyboroxin
CAS Number: 102-24-9
MDL Number: MFCD00005946
Molecular Formula: C3H9B3O6
Molecular Weight: 173.53
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 9
Specific Gravity (20/20): 1.22
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Trimethoxycyclotriboroxane
Formula:
C₃H₉B₃O₆ MW: 173.53 g/mol Density: 1.22 (20 °C) Flash Pt: 9 °C |
MDL Number:
MFCD00005946 CAS Number: 102-24-9 |
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...