(Trimethylsilyl)methyl trifluoromethanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: (Trimethylsilyl)methyl Triflate, Trifluoromethanesulphonic Acid (Trimethylsilyl)methyl Ester

T1579-5G T1579-1G T1579-25G
TCT1579-005GEA 195.6 USD
TCT1579-005G TCT1579-1G TCT1579-25G
(Trimethylsilyl)methyl trifluoromethanesulfonate ≥98.0%
(Trimethylsilyl)methyl trifluoromethanesulfonate

[Trimethylsilylmethylating Reagent]
CAS Number: 64035-64-9
MDL Number: MFCD00009914
Molecular Formula: C5H11F3O3SSi
Molecular Weight: 236.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 168
Flash Point (°C): 48
Specific Gravity (20/20): 1.19


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid (Trimethylsilyl)methyl Ester, (Trimethylsilyl)methyl Triflate

Formula: C₅H₁₁F₃O₃SSi
MW: 236.28 g/mol
Boiling Pt: 168 °C
Density: 1.19 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00009914
CAS Number: 64035-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=863215f4269d2671bc0dd991f68f419f5296f771&vid=863215f4269d2671bc0dd991f68f419f5296f771&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR