Tris(2-cyanoethyl)amine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

T1577-5G T1577-25G
TCT1577-005GEA 55 USD
TCT1577-005G TCT1577-25G
Tris(2-cyanoethyl)amine ≥99.0% (by titrimetric analysis)
Tris(2-cyanoethyl)amine

CAS Number: 7528-78-1
MDL Number: MFCD00671575
Molecular Formula: C9H12N4
Molecular Weight: 176.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 140
Melting point (°C): 60


Other Product Names / Synonyms: 3,3',3''-Nitrilotripropionitrile

Formula: C₉H₁₂N₄
Boiling Pt: 140 °C
Melting Pt: 60 °C
MDL Number: MFCD00671575
CAS Number: 7528-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cfd145c654600928bd144907775b8c1ad1bf8c1&vid=5cfd145c654600928bd144907775b8c1ad1bf8c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR