Tetramethylammonium triacetoxyborohydride ≥95.0%
Supplier: TCI America
T1553-5G
T1553-25G
TCT1553-005GEA
125
USD
TCT1553-005G
TCT1553-25G
Tetramethylammonium triacetoxyborohydride ≥95.0%
Tetramethylammonium triacetoxyborohydride
CAS Number: 109704-53-2
MDL Number: MFCD00012196
Molecular Formula: C10H22BNO6
Molecular Weight: 263.10
Purity/Analysis Method:
Formula:
C₁₀H₂₂BNO₆ |
MDL Number:
MFCD00012196 CAS Number: 109704-53-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...