Tris(3-aminopropyl)amine ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N1,N1-Bis(3-aminopropyl)propane-1,3-diamine, 3,3',3''-Nitrilotris(propylamine)

T1540-5ML T1540-25ML
TCT1540-005MLEA 260.5 USD
TCT1540-005ML TCT1540-25ML
Tris(3-aminopropyl)amine ≥95.0% (by GC, titration analysis)
Tris(3-aminopropyl)amine

CAS Number: 4963-47-7
MDL Number: MFCD00191593
Molecular Formula: C9H24N4
Molecular Weight: 188.32
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 150
Specific Gravity (20/20): 0.95
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3,3',3''-Nitrilotris(propylamine)

Formula: C₉H₂₄N₄
MW: 188.32 g/mol
Boiling Pt: 150 °C
Density: 0.95 (20 °C)
MDL Number: MFCD00191593
CAS Number: 4963-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2d4a34582623f1032e0d3bb57583513d970556&vid=fe2d4a34582623f1032e0d3bb57583513d970556&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR