2-(Trimethylsilyloxy)furan ≥95.0%
Supplier: TCI America
Synonyms:
(2-Furanyloxy)trimethylsilane
T1536-5G
T1536-25G
TCT1536-005GEA
281
USD
TCT1536-005G
TCT1536-25G
2-(Trimethylsilyloxy)furan ≥95.0%
2-(Trimethylsilyloxy)furan
CAS Number: 61550-02-5
MDL Number: MFCD00011729
Molecular Formula: C7H12O2Si
Molecular Weight: 156.26
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 50
Flash Point (°C): 18
Specific Gravity (20/20): 0.95
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (2-Furanyloxy)trimethylsilane
Formula:
C₇H₁₂O₂Si Boiling Pt: 50 °C Density: 0.95 (20 °C) Flash Pt: 18 °C |
MDL Number:
MFCD00011729 CAS Number: 61550-02-5 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...