(3-Trimethylsilyl-2-propynyl)triphenylphosphonium bromide ≥98.0%

Supplier: TCI America

T1498-1G T1498-5G
TCT1498-1GEA 85 USD
TCT1498-1G TCT1498-005G
(3-Trimethylsilyl-2-propynyl)triphenylphosphonium bromide ≥98.0%
(3-Trimethylsilyl-2-propynyl)triphenylphosphonium bromide

CAS Number: 42134-49-6
MDL Number: MFCD00012029
Molecular Formula: C24H26BrPSi
Molecular Weight: 453.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow

Formula: C₂₄H₂₆BrPSi
MW: 453.43 g/mol
MDL Number: MFCD00012029
CAS Number: 42134-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94cb20dd29f05991cba7350c72b00bea9e1bc8af&vid=94cb20dd29f05991cba7350c72b00bea9e1bc8af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR