2-(Trimethylsiloxy)-1,3-butadiene ≥95.0%
Supplier: TCI America
Synonyms:
(Buta-1,3-dien-2-yloxy)trimethylsilane
T1469-5ML
T1469-25ML
TCT1469-005MLEA
93.76
USD
TCT1469-005ML
TCT1469-25ML
2-(Trimethylsiloxy)-1,3-butadiene ≥95.0%
2-(Trimethylsiloxy)-1,3-butadiene
CAS Number: 38053-91-7
MDL Number: MFCD00009852
Molecular Formula: C7H14OSi
Molecular Weight: 142.27
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 55
Flash Point (°C): 12
Specific Gravity (20/20): 0.82
Storage Temperature: 0-10°C
Formula:
C₇H₁₄OSi MW: 142.27 g/mol Boiling Pt: 55 °C Density: 0.82 (20 °C) Flash Pt: 12 °C |
MDL Number:
MFCD00009852 CAS Number: 38053-91-7 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...