1,5,6,7-Tetrahydro-4H-indol-4-one ≥97.0%

Supplier: TCI America

Synonyms: 4-Oxo-4,5,6,7-tetrahydroindole, 6,7-Dihydro-1H-indol-4(5H)-one

T1435-25G T1435-5G
TCT1435-025GEA 90 USD
TCT1435-025G TCT1435-005G
1,5,6,7-Tetrahydro-4H-indol-4-one ≥97.0%
1,5,6,7-Tetrahydro-4H-indol-4-one

CAS Number: 13754-86-4
MDL Number: MFCD00075438
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 190


Other Product Names / Synonyms: 4-Oxo-4,5,6,7-tetrahydroindole

Formula: C₈H₉NO
MW: 135.17 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00075438
CAS Number: 13754-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93df55ea77ac106e67b8fbbe0d272ef79b8031d9&vid=93df55ea77ac106e67b8fbbe0d272ef79b8031d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR