Tetrapentylammonium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Tetraamylammonium chloride, Tetrapentylammonium chloride
T1433-5G
T1433-25G
TCT1433-005GEA
149.62
USD
TCT1433-005G
TCT1433-25G
Tetrapentylammonium chloride ≥98.0% (by titrimetric analysis)
Tetrapentylammonium chloride
CAS Number: 4965-17-7
MDL Number: MFCD00011857
Molecular Formula: C20H44ClN
Molecular Weight: 334.03
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Other Product Names / Synonyms: Tetrapentylammonium Chloride
Formula:
C₂₀H₄₄ClN MW: 334.03 g/mol Melting Pt: 139.85 °C |
MDL Number:
MFCD00011857 CAS Number: 4965-17-7 EINECS: 225-606-4 UN: 2811 ADR: 6.1,I |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...