Tetrapentylammonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Tetraamylammonium bromide

T1432-25G T1432-5G
TCT1432-025GEA 112.9 USD
TCT1432-025G TCT1432-005G
Tetrapentylammonium bromide ≥98.0% (by titrimetric analysis)
Tetrapentylammonium bromide

CAS Number: 866-97-7
MDL Number: MFCD00011856
Molecular Formula: C20H44BrN
Molecular Weight: 378.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 101


Other Product Names / Synonyms: Tetrapentylammonium Bromide

Formula: C₂₀H₄₄BrN
MW: 378.48 g/mol
Melting Pt: 101 °C
Density: 1.0662 g/cm³ (25 °C)
MDL Number: MFCD00011856
CAS Number: 866-97-7
EINECS: 212-756-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02910929365518e83f478368d737dcfbf5931dc5&vid=02910929365518e83f478368d737dcfbf5931dc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR