4-Methyl-2,3,5,6-tetrabromophenol ≥95.0%

Supplier: TCI America

Synonyms: 2,3,5,6-Tetrabromo-4-methylphenol, 2,3,5,6-Tetrabromo-p-cresol, 4-Hydroxy-2,3,5,6-tetrabromotoluene

T1430-25G T1430-5G
TCT1430-25GEA 148.6 USD
TCT1430-25G TCT1430-005G
4-Methyl-2,3,5,6-tetrabromophenol ≥95.0%
4-Methyl-2,3,5,6-tetrabromophenol

CAS Number: 37721-75-8
MDL Number: MFCD00060318
Molecular Formula: C7H4Br4O
Molecular Weight: 423.72
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 198


Other Product Names / Synonyms: 2,3,5,6-Tetrabromo-4-methylphenol

Formula: C₇H₄Br₄O
MW: 423.72 g/mol
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 198 °C
MDL Number: MFCD00060318
CAS Number: 37721-75-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e37029f6ceb447d15ba6d658d0b060501e8eeda&vid=2e37029f6ceb447d15ba6d658d0b060501e8eeda&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR