Trilinolein ≥95.0%

Supplier: TCI America

Synonyms: 1,2,3-Tri(cis,cis-9,12-octadecadienoyl) Glycerol, Glycerol Tri-9,12-octadecadienoate

T1388-25G T1388-5G
TCT1388-25GEA 920 USD
TCT1388-25G TCT1388-005G
Trilinolein ≥95.0%
Trilinolein

CAS Number: 537-40-6
MDL Number: MFCD00042909
Molecular Formula: C57H98O6
Molecular Weight: 879.41
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Tri-9,12-octadecadienoate, 1,2,3-Tri(cis,cis-9,12-octadecadienoyl) Glycerol

Formula: C₅₇H₉₈O₆
MW: 879.41 g/mol
Density: 0.92 (20 °C)
MDL Number: MFCD00042909
CAS Number: 537-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=177b8976826b5b64f89fd71d2c06fc6bb990904e&vid=177b8976826b5b64f89fd71d2c06fc6bb990904e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR