Methyltri-n-octylammonium chloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tri-n-octylmethylammonium Chloride, N-Methyl-N,N-dioctyloctan-1-aminium chloride

T1365-25G T1365-500G
TCT1365-025GEA 107 USD
TCT1365-025G TCT1365-500G
Methyltri-n-octylammonium chloride ≥95.0% (by titrimetric analysis)
Methyltri-n-octylammonium chloride

CAS Number: 5137-55-3
MDL Number: MFCD00011862
Molecular Formula: C25H54ClN
Molecular Weight: 404.16
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Flash Point (°C): 102
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Tri-n-octylmethylammonium Chloride

Formula: C₂₅H₅₄ClN
MW: 404.16 g/mol
Flash Pt: 102 °C
MDL Number: MFCD00011862
CAS Number: 5137-55-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2eb71f655d1eebd5f8befcf45e938d124e7e1d0e&vid=a97bd16303f549c4d3af9bd4284e367b729ef33d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR