1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1356-5G T1356-25G
TCT1356-005GEA 34 USD
TCT1356-005G TCT1356-025G
1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride

CAS Number: 43064-12-6
MDL Number: MFCD00012752
Molecular Formula: C11H13N
Molecular Weight: 195.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 203


Other Product Names / Synonyms: 4-Phenyl-1,2,3,6-tetrahydropyridine Hydrochloride

Formula: C₁₁H₁₃N
Melting Pt: 203 °C
MDL Number: MFCD00012752
CAS Number: 43064-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2799e7b260f476b2cebde7fcf03be472312a2c8d&vid=2799e7b260f476b2cebde7fcf03be472312a2c8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR