1,3,5-Triacetylbenzene ≥98.0%

Supplier: TCI America

T1337-5G T1337-25G
TCT1337-005GEA 207.81 USD
TCT1337-005G TCT1337-025G
1,3,5-Triacetylbenzene ≥98.0%
1,3,5-Triacetylbenzene

CAS Number: 779-90-8
MDL Number: MFCD00008741
Molecular Formula: C12H12O3
Molecular Weight: 204.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 163

Formula: C₁₂H₁₂O₃
Melting Pt: 163 °C
MDL Number: MFCD00008741
CAS Number: 779-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2675d43313bf14c2c16e2e0a38c989a6421f71ac&vid=2675d43313bf14c2c16e2e0a38c989a6421f71ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR