3-Trifluoromethyl-4'-methoxydiphenylamine

Supplier: TCI America

T1326-1G T1326-5G
TCT1326-001GEA 99 USD
TCT1326-001G TCT1326-005G
3-Trifluoromethyl-4'-methoxydiphenylamine
3-Trifluoromethyl-4'-methoxydiphenylamine

CAS Number: 1494-26-4
MDL Number: MFCD00143198
Molecular Formula: C14H12F3NO
Molecular Weight: 267.25
Form: Crystal
Color: White
Melting point (°C): 58


Other Product Names / Synonyms: N-(3-Trifluoromethylphenyl)-p-anisidine

Formula: C₁₄H₁₂F₃NO
Melting Pt: 58 °C
MDL Number: MFCD00143198
CAS Number: 1494-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa76dbad2daf13d631d8c3af3dfa400f5e3faa02&vid=aa76dbad2daf13d631d8c3af3dfa400f5e3faa02&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR