Tetrabutylammonium bifluoride ≥95.0%
Supplier: TCI America
Synonyms:
Tetrabutylammonium Hydrogen Difluoride
T1295-5G
T1295-25G
TCT1295-005GEA
135.32
USD
TCT1295-005G
TCT1295-25G
Tetrabutylammonium bifluoride ≥95.0%
Tetrabutylammonium bifluoride
CAS Number: 23868-34-0
MDL Number: MFCD00077877
Molecular Formula: C16H37F2N
Molecular Weight: 281.48
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Color: Colorless
Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: Tetrabutylammonium Hydrogen Difluoride
Formula:
C₁₆H₃₇F₂N MW: 281.48 g/mol |
MDL Number:
MFCD00077877 CAS Number: 23868-34-0 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...