N-(Trimethylsilyl)morpholine ≥97.0%
Supplier: TCI America
T1277-5ML
T1277-25ML
TCT1277-005MLEA
58.6
USD
TCT1277-005ML
TCT1277-025ML
N-(Trimethylsilyl)morpholine ≥97.0%
N-(Trimethylsilyl)morpholine
CAS Number: 13368-42-8
MDL Number: MFCD00010098
Molecular Formula: C7H17NOSi
Molecular Weight: 159.30
Purity/Analysis Method:
Formula:
C₇H₁₇NOSi Boiling Pt: 102 °C Density: 0.90 (20 °C) Flash Pt: 29 °C |
MDL Number:
MFCD00010098 CAS Number: 13368-42-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...