1,1,1-Tris(4-hydroxyphenyl)ethane ≥98.0%

Supplier: TCI America

Synonyms: 4,4',4''-(Ethane-1,1,1-triyl)triphenol

T1254-25G T1254-500G T1254-100G
TCT1254-025GEA 44 USD
TCT1254-025G TCT1254-500G TCT1254-100G
1,1,1-Tris(4-hydroxyphenyl)ethane ≥98.0%
1,1,1-Tris(4-hydroxyphenyl)ethane

CAS Number: 27955-94-8
MDL Number: MFCD00012180
Molecular Formula: C20H18O3
Molecular Weight: 306.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 248

Formula: C₂₀H₁₈O₃
MW: 306.36 g/mol
Melting Pt: 248 °C
MDL Number: MFCD00012180
CAS Number: 27955-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caa433415b51ca61559c999839f735b8fbb2fae7&vid=caa433415b51ca61559c999839f735b8fbb2fae7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR