Potassium-p-toluenethiosulfonate ≥98.0% (by HPLC)

Supplier: TCI America

T1185-5G T1185-25G
TCT1185-005GEA 48 USD
TCT1185-005G TCT1185-025G
Potassium-p-toluenethiosulfonate ≥98.0% (by HPLC)
Potassium-p-toluenethiosulfonate

CAS Number: 28519-50-8
MDL Number: MFCD00003547
Molecular Formula: C7H8O2S2
Molecular Weight: 226.35
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 229


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenethiosulfonic Acid Potassium Salt

Formula: C₇H₈O₂S₂
Melting Pt: 229 °C
MDL Number: MFCD00003547
CAS Number: 28519-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cdc7a62265d3c47a8e02856dcb72f8cbe74968b&vid=2cdc7a62265d3c47a8e02856dcb72f8cbe74968b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR