Trimethylsilylmethyl azide ≥97.0%

Supplier: TCI America

T1184-1G T1184-5G
TCT1184-001GEA 159.73 USD
TCT1184-001G TCT1184-5G
Trimethylsilylmethyl azide ≥97.0%
Trimethylsilylmethyl azide

CAS Number: 87576-94-1
MDL Number: MFCD00059964
Molecular Formula: C4H11N3Si
Molecular Weight: 129.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Flash Point (°C): 6
Specific Gravity (20/20): 0.87


Product Categories: Synthetic Organic Chemistry, Amination, Silicon Compounds (for Synthesis), Azide Compounds (Click Chemistry)
Other Product Names / Synonyms: Azidomethyltrimethylsilane

Formula: C₄H₁₁N₃Si
Boiling Pt: 60 °C
Density: 0.87 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00059964
CAS Number: 87576-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=370b8cbf7848b486ddf08414cea5340f1bdf6851&vid=370b8cbf7848b486ddf08414cea5340f1bdf6851&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR