Tripentylamine ≥98.0%
Supplier: TCI America
T1177-25ML
T1177-500ML
TCT1177-025MLEA
36
USD
TCT1177-025ML
TCT1177-500ML
Tripentylamine ≥98.0%
Tripentylamine
CAS Number: 621-77-2
MDL Number: MFCD00015270
Molecular Formula: C15H33N
Molecular Weight: 227.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 240
Flash Point (°C): 81
Specific Gravity (20/20): 0.79
Other Product Names / Synonyms: Tripentylamine
Formula:
C₁₅H₃₃N Boiling Pt: 240 °C Density: 0.79 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00015270 CAS Number: 621-77-2 EINECS: 210-705-7 UN: 2735 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...