Trimethylhexamethylene Diisocyanate (2,2,4- and 2,4,4- mixture) ≥97.0%
Supplier: TCI America
CAS Number: 28679-16-5
MDL Number: MFCD00040422
Molecular Formula: C11H18N2O2
Molecular Weight: 210.28
Purity/Analysis Method:
Formula:
C₁₁H₁₈N₂O₂ Density: 1,02 (20 °C) |
MDL Number:
MFCD00040422 CAS Number: 28679-16-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...