2,6-Toluenediisocyanate ≥98.0%

Supplier: TCI America

Synonyms: 2,6-TDI, 2-Methyl-m-phenylene diisocyanate, Tolylene 2,6-diisocyanate

T1153-5G T1153-1G T1153-25G
TCT1153-005GEA 126 USD
TCT1153-005G TCT1153-001G TCT1153-25G
2,6-Toluenediisocyanate ≥98.0%
2,6-Toluenediisocyanate

CAS Number: 91-08-7
MDL Number: MFCD00002010
Molecular Formula: C9H6N2O2
Molecular Weight: 174.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 103
Flash Point (°C): 127
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 2-Methyl-m-phenylene Diisocyanate, Toluene-2,6-diisocyanate, 2-Methyl-1,3-phenylene Diisocyanate, 2,6-Diisocyanatotoluene

Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Boiling Pt: 103 °C
Density: 1.23 (20 °C)
Flash Pt: 127 °C
MDL Number: MFCD00002010
CAS Number: 91-08-7
EINECS: 202-039-0
UN: 2078
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88281c324407a8c32b17a3dae614c5ad7dde6186&vid=88281c324407a8c32b17a3dae614c5ad7dde6186&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR