Tetraethyl orthocarbonate ≥97.0%

Supplier: TCI America

T1107-5ML T1107-25ML
TCT1107-005MLEA 46 USD
TCT1107-005ML TCT1107-025ML
Tetraethyl orthocarbonate ≥97.0%
Tetraethyl orthocarbonate

CAS Number: 78-09-1
MDL Number: MFCD00009221
Molecular Formula: C9H20O4
Molecular Weight: 192.26
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 160
Flash Point (°C): 52
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Tetraethyl Orthocarbonate

Formula: C₉H₂₀O₄
MW: 192.26 g/mol
Boiling Pt: 160 °C
Density: 0.92 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00009221
CAS Number: 78-09-1
EINECS: 201-082-2
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd256cedd2772a0684592026a78b1bb0bc29b828&vid=bd256cedd2772a0684592026a78b1bb0bc29b828&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR