2,4,6-T (2,4,6-trichlorophenoxy acetic acid) ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T1063-5G
T1063-25G
TCT1063-005GEA
70.1
USD
TCT1063-005G
TCT1063-25G
2,4,6-T (2,4,6-trichlorophenoxy acetic acid) ≥98.0% (by GC, titration analysis)
2,4,6-T (2,4,6-trichlorophenoxy acetic acid)
CAS Number: 575-89-3
MDL Number: MFCD00053498
Molecular Formula: C8H5Cl3O3
Molecular Weight: 255.48
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 185
Formula:
C₈H₅Cl₃O₃ Melting Pt: 185 °C |
MDL Number:
MFCD00053498 CAS Number: 575-89-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...