2-Butynoic acid methyl ester ≥97.0%

Supplier: TCI America

Synonyms: Methyl 2-butynoate, Methyl but-2-ynoate

T1053-1ML T1053-5ML T1053-25ML
TCT1053-001MLEA 56 USD
TCT1053-001ML TCT1053-005ML TCT1053-25ML
2-Butynoic acid methyl ester ≥97.0%
2-Butynoic acid methyl ester

CAS Number: 23326-27-4
MDL Number: MFCD00009273
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 148
Flash Point (°C): 45
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2-Butynoic Acid Methyl Ester, Methyl 2-Butynoate, Tetrolic Acid Methyl Ester

Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 148 °C
Density: 1.00 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00009273
CAS Number: 23326-27-4
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19da5306cfd802801227b6126b002991d9815a39&vid=19da5306cfd802801227b6126b002991d9815a39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR