Tetramethyl orthocarbonate ≥97.0%

Supplier: TCI America

Synonyms: Tetramethylorthocarbonate

T1045-5G T1045-25G
TCT1045-005GEA 250 USD
TCT1045-005G TCT1045-25G
Tetramethyl orthocarbonate ≥97.0%
Tetramethyl orthocarbonate

CAS Number: 1850-14-2
MDL Number: MFCD00008473
Molecular Formula: C5H12O4
Molecular Weight: 136.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 114
Flash Point (°C): 22
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Tetramethyl Orthocarbonate

Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 114 °C
Density: 1.02 (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00008473
CAS Number: 1850-14-2
EINECS: 217-438-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dcc849bed61766b8ec4c79700a15b48b6fc0404&vid=5dcc849bed61766b8ec4c79700a15b48b6fc0404&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR