3,4,4'-Trichlorocarbanilide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 3,4,4'-Trichlorodiphenylurea, 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea, Triclocarban

T1015-25G T1015-500G
TCT1015-025GEA 50.4 USD
TCT1015-025G TCT1015-500G
3,4,4'-Trichlorocarbanilide ≥98.0% (by HPLC, total nitrogen)
3,4,4'-Trichlorocarbanilide

CAS Number: 101-20-2
MDL Number: MFCD00013254
Molecular Formula: C13H9Cl3N2O
Molecular Weight: 315.58
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Ureas (Multi-component Reaction)
Other Product Names / Synonyms: 3,4,4'-Trichlorodiphenylurea, Triclocarban, 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea

Formula: C₁₃H₉Cl₃N₂O
MW: 315.58 g/mol
MDL Number: MFCD00013254
CAS Number: 101-20-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb01e5f17bf016c4d79e1843499e9a4178cd87f9&vid=bb01e5f17bf016c4d79e1843499e9a4178cd87f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR