Triethyl orthovalerate ≥95.0%

Supplier: TCI America

Synonyms: 1,1,1-Triethoxypentane, Orthovaleric Acid Triethyl Ester

T0977-250ML T0977-25ML
TCT0977-250MLEA 896.6 USD
TCT0977-250ML TCT0977-025ML
Triethyl orthovalerate ≥95.0%
Triethyl orthovalerate

CAS Number: 919-29-9
MDL Number: MFCD00059383
Molecular Formula: C11H24O3
Molecular Weight: 204.31
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 50
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: 1,1,1-Triethoxypentane, Orthovaleric Acid Triethyl Ester

Formula: C₁₁H₂₄O₃
MW: 204.31 g/mol
Boiling Pt: 50 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00059383
CAS Number: 919-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=533c740176a768451bf307671ca2eea0b8be92e9&vid=533c740176a768451bf307671ca2eea0b8be92e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR