Tetrahydrofuran-2,3,4,5-tetracarboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T0975-500G
T0975-25G
TCT0975-500GEA
1256
USD
TCT0975-500G
TCT0975-025G
Tetrahydrofuran-2,3,4,5-tetracarboxylic acid ≥98.0% (by titrimetric analysis)
Tetrahydrofuran-2,3,4,5-tetracarboxylic acid
CAS Number: 26106-63-8
MDL Number: MFCD00005364
Molecular Formula: C8H8O9
Molecular Weight: 248.14
Purity/Analysis Method:
Formula:
C₈H₈O₉ Melting Pt: 205 °C |
MDL Number:
MFCD00005364 CAS Number: 26106-63-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...