N,N,N',N'-Tetramethyl-1,2-diaminopropane ≥92.0%

Supplier: TCI America

Synonyms: N,N,N',N'-Tetramethyl-1,2-propanediamine, 1,2-Bis(dimethylamino)propane

T0947-25ML T0947-5ML
TCT0947-025MLEA 252.1 USD
TCT0947-025ML TCT0947-5ML
N,N,N',N'-Tetramethyl-1,2-diaminopropane ≥92.0%
N,N,N',N'-Tetramethyl-1,2-diaminopropane

CAS Number: 1822-45-3
MDL Number: MFCD00014871
Molecular Formula: C7H18N2
Molecular Weight: 130.24
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 72
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: 1,2-Bis(dimethylamino)propane, N,N,N',N'-Tetramethyl-1,2-propanediamine

Formula: C₇H₁₈N₂
MW: 130.24 g/mol
Boiling Pt: 72 °C
Density: 0.81 (20 °C)
MDL Number: MFCD00014871
CAS Number: 1822-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85e452ff4c1a7c8254a90e390d341edf2cd9e1aa&vid=85e452ff4c1a7c8254a90e390d341edf2cd9e1aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >92.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR