Tridecylamine (=Monotridecylamine) (mixture of branched chain isomers) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0936-25ML T0936-500ML
TCT0936-025MLEA 24 USD
TCT0936-025ML TCT0936-500ML
Tridecylamine (=Monotridecylamine) (mixture of branched chain isomers) ≥98.0% (by titrimetric analysis)
Tridecylamine (=monotridecylamine) (mixture of branched chain isomers)

CAS Number: 86089-17-0 MDL Number: MFCD00008156 Molecular Formula: C13H29N Molecular Weight: 199.38 Purity/Analysis Method: 98.0% (T) Form: Clear Liquid Color: Colorless Flash Point (°C): 110 Specific Gravity (20/20): 0.82

Formula: C₁₃H₂₉N
Density: 0.82 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008156
CAS Number: 86089-17-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR