Tamarind gum (from tamarind seed, polysaccharide)

Supplier: TCI America

T0909-500G T0909-25G
TCT0909-500GEA 154.32 USD
TCT0909-500G TCT0909-025G
Tamarind gum (from tamarind seed, polysaccharide)
Tamarind gum

Form: Crystal
Color: White

MDL Number: MFCD00148561
CAS Number: 39386-78-2

Order Now

Specification Test Results

Identification test(1) to pass test
Identification test(2) to pass test
Arsenic max. 2.0ppm(as As2O3)
Heavy metals max. 20ppm(as Pb)
Protein max. 3.0%
Drying loss max. 7.0 %
Ash Content max. 5.0 %
Starch to pass test
Diethyl ether soluble matter max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR