Butyl mercaptoacetate ≥98.0%

Supplier: TCI America

Synonyms: Butyl Mercaptoacetate, Butyl Thioglycolate, Mercaptoacetic acid butyl ester, Thioglycolic acid butyl ester

T0903-500ML T0903-25ML
TCT0903-500MLEA 286.6 USD
TCT0903-500ML TCT0903-025ML
Butyl mercaptoacetate ≥98.0%
Butyl mercaptoacetate

CAS Number: 10047-28-6
MDL Number: MFCD00022087
Molecular Formula: C6H12O2S
Molecular Weight: 148.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 194
Flash Point (°C): 66
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Thioglycolic Acid Butyl Ester, Mercaptoacetic Acid Butyl Ester, Butyl Mercaptoacetate

Formula: C₆H₁₂O₂S
MW: 148.23 g/mol
Boiling Pt: 194 °C
Density: 1.03 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00022087
CAS Number: 10047-28-6
EINECS: 233-156-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2795db80c7dbe6e1d9045188880865fcb0e8f616&vid=2795db80c7dbe6e1d9045188880865fcb0e8f616&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR