Bithionol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Bithionol, Bis(3,5-dichloro-2-hydroxyphenyl) sulphide, 2,2'-Thiobis(4,6-dichlorophenol), 6,6'-Thiobis(2,4-dichlorophenol)
CAS Number: 97-18-7
MDL Number: MFCD00055727
Molecular Formula: C12H6Cl4O2S
Molecular Weight: 356.04
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 187
Product Categories: Materials Chemistry, Monomers, Diphenyl Sulfides (for High-Performance Polymer Research), Bioscience, Anthelmintic Ingredients
Other Product Names / Synonyms: Bis(3,5-dichloro-2-hydroxyphenyl) Sulfide, Bithionol
Formula:
C₁₂H₆Cl₄O₂S MW: 356.0518 g/mol Melting Pt: 187 °C |
MDL Number:
MFCD00055727 CAS Number: 97-18-7 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...