Methyltrialkylammonium chloride (mixture of C8-C10)

Supplier: TCI America

T0826-500G T0826-25G T0826-100G
TCT0826-500GEA 98.32 USD
TCT0826-500G TCT0826-025G TCT0826-100G
Methyltrialkylammonium chloride (mixture of C8-C10)
Methyltrialkylammonium chloride (mixture of C8-C10)

[Phase-transfer Catalyst]
CAS Number: 63393-96-4
MDL Number: MFCD00011862
Molecular Formula: C25H54ClN
Form: Viscous Clear Liquid
Color: Colorless
Flash Point (°C): 132


Other Product Names / Synonyms: Methyltrioctylammonium Chloride, Tricaprylmethylammonium Chloride

Formula: CH₃N(C₈-C₁₀n-Alkyl)₃Cl
Flash Pt: 132 °C
MDL Number: MFCD00011862
CAS Number: 63393-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR