2-Amino-4-chloro-6-nitrophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 6358-08-3
MDL Number: MFCD00035925
Molecular Formula: C6H5ClN2O3
Molecular Weight: 188.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 152
Other Product Names / Synonyms: 5-Chloro-2-hydroxy-3-nitroaniline
Formula:
C₆H₅ClN₂O₃ Melting Pt: 152 °C |
MDL Number:
MFCD00035925 CAS Number: 6358-08-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...