Nɑ-Tosyl-L-arginine
Supplier: TCI America
T0684-5G
T0684-1G
TCT0684-005GEA
37
USD
TCT0684-005G
TCT0684-1G
Nɑ-Tosyl-L-arginine
Nalpha-Tosyl-L-arginine
CAS Number: 1159-15-5
MDL Number: MFCD00019732
Molecular Formula: C13H20N4O4S
Molecular Weight: 328.39
Form: Crystal
Color: White
Other Product Names / Synonyms: Nalpha-p-Toluenesulfonyl-L-arginine, Tos-Arg-OH
Formula:
C₁₃H₂₀N₄O₄S |
MDL Number:
MFCD00019732 CAS Number: 1159-15-5 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (after drying) |
Specific rotation [a]20/D | -14.0 to -16.0 deg (C=2, 1mol/L HCl) (after drying) |
Drying loss | 10.0 to 16.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...