Thiopropionamide ≥98.0%
Supplier: TCI America
T0669-10G
T0669-25G
TCT0669-010GEA
157
USD
TCT0669-010G
TCT0669-25G
Thiopropionamide ≥98.0%
Thiopropionamide
CAS Number: 631-58-3
MDL Number: MFCD00059864
Molecular Formula: C3H7NS
Molecular Weight: 89.16
Purity/Analysis Method:
Formula:
C₃H₇NS Melting Pt: 40 °C |
MDL Number:
MFCD00059864 CAS Number: 631-58-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...