Tecnazene ≥98.0%

Supplier: TCI America
Warning

Synonyms: Tecnazene, 2,3,5,6-Tetrachloronitrobenzene

T0624-25G T0624-500G
TCT0624-025GEA 102.69 USD
TCT0624-025G TCT0624-500G
Tecnazene ≥98.0%
Tecnazene

CAS Number: 117-18-0
MDL Number: MFCD00007066
Molecular Formula: C6HCl4NO2
Molecular Weight: 260.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 100

Formula: C₆HCl₄NO₂
MW: 260.89 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 100 °C
Density: 1.744 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007066
CAS Number: 117-18-0
EINECS: 204-178-2
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e7c4ff035bbb8f4359df5b8bdb33578b9ce2576&vid=de9a01bb61b11d81d0ab7900ca783d53ef651807&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR