(±)-3,5,5-Trimethyl-1-hexanol ≥85.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3,5,5-Trimethyl-1-hexanol, 3,5,5-Trimethylhexan-1-ol, 3,5,5-Trimethylhexyl alcohol
T0596-25ML
T0596-500ML
TCT0596-025MLEA
22.39
USD
TCT0596-025ML
TCT0596-500ML
(±)-3,5,5-Trimethyl-1-hexanol ≥85.0%
(±)-3,5,5-Trimethyl-1-hexanol
CAS Number: 3452-97-9
MDL Number: MFCD00036138
Molecular Formula: C9H20O
Molecular Weight: 144.26
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 193
Flash Point (°C): 79
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: 3,5,5-Trimethylhexyl Alcohol
Formula:
(CH₃)₃CCH₂CH(CH₃)CH₂CH₂OH MW: 144.26 g/mol Boiling Pt: 193 °C Melting Pt: –70 °C Density: 0.83 (20 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036138 CAS Number: 3452-97-9 EINECS: 222-376-7 UN: 3082 ADR: 9 III |
|
Specification Test Results
Purity/Analysis Method | >85.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...