5-Chloro-2-(2-chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2224-5G A2224-25G
TCA2224-5GEA 130.8 USD
TCA2224-5G TCA2224-25G
5-Chloro-2-(2-chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)
5-Chloro-2-(2-chlorophenoxy)aniline

CAS Number: 56966-48-4
MDL Number: MFCD00025218
Molecular Formula: C12H9Cl2NO
Molecular Weight: 254.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 210
Specific Gravity (20/20): 1.35


Other Product Names / Synonyms: 5-Chloro-2-(2-chlorophenoxy)aniline

Formula: C₁₂H₉Cl₂NO
Boiling Pt: 210 °C
Density: 1.35 (20 °C)
MDL Number: MFCD00025218
CAS Number: 56966-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2105831024d97427a4ea6e355bdc6c58d6fe7633&vid=2105831024d97427a4ea6e355bdc6c58d6fe7633&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR