Trimethylene sulfide ≥98.0%

Supplier: TCI America

T0474-25ML T0474-5ML
TCT0474-025MLEA 450 USD
TCT0474-025ML TCT0474-005ML
Trimethylene sulfide ≥98.0%
Trimethylene sulfide

CAS Number: 287-27-4
MDL Number: MFCD00042751
Molecular Formula: C3H6S
Molecular Weight: 74.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 94
Flash Point (°C): -2
Specific Gravity (20/20): 1.03

Formula: C₃H₆S
MW: 74.14 g/mol
Boiling Pt: 94 °C
Density: 1.03 (20 °C)
Flash Pt: –2 °C
MDL Number: MFCD00042751
CAS Number: 287-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d5d84ac90c4facf1b43fb5c7f3fb9ac361e5213&vid=8d5d84ac90c4facf1b43fb5c7f3fb9ac361e5213&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR