2-[(Acetylthio)methyl]-3-phenylpropionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A2220-1G A2220-5G
TCA2220-1GEA 62 USD
TCA2220-1G TCA2220-5G
2-[(Acetylthio)methyl]-3-phenylpropionic acid ≥98.0% (by titrimetric analysis)
2-[(Acetylthio)methyl]-3-phenylpropionic acid

CAS Number: 91702-98-6
MDL Number: MFCD00236751
Molecular Formula: C12H14O3S
Molecular Weight: 238.30
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid

Formula: C₁₂H₁₄O₃S
MDL Number: MFCD00236751
CAS Number: 91702-98-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fea09fe07b5ae09e2a348c761745d2d22460456&vid=8fea09fe07b5ae09e2a348c761745d2d22460456&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR